Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHI3L2
Entrez Gene ID
1117
Gene Name
chitinase 3 like 2
Gene Aliases
CHIL2, YKL-39, YKL39
Chromosome Location
Chr.1: 111227659 - 111243440 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025197.1 NP_001020368.1 7 - 8 959 104
NM_001025199.1 NP_001020370.1 7 - 8 1019 104
NM_004000.2 NP_003991.2 8 - 9 989 104
GenBank mRNA AK298466.1 - 7 - 8 910 104
BC011460.1 - 8 - 9 927 104
BT006767.1 - 8 - 9 918 104
U49835.1 - 8 - 9 938 104
U58514.1 - 8 - 9 954 104
U58515.1 - 7 - 8 1020 104

View Full Product Details