Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD9L
Entrez Gene ID
219285
Gene Name
sterile alpha motif domain containing 9 like
Gene Aliases
C7orf6, DRIF2, UEF1
Chromosome Location
Chr.7: 93130054 - 93148401 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303496.1 NP_001290425.1 2 - 3 256 114
XM_005250193.3 XP_005250250.1 2 - 3 269 114
GenBank mRNA AF474973.1 - 2 - 3 214 114
AY195585.1 - 2 - 3 213 114

View Full Product Details