The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303497.1 | NP_001290426.1 | 3 - 4 | 590 | 61 |
NM_001303498.1 | NP_001290427.1 | 2 - 3 | 326 | 61 | |
XM_005250193.3 | XP_005250250.1 | 3 - 4 | 398 | 61 | |
GenBank mRNA | AF474973.1 | - | 3 - 4 | 343 | 61 |
AY195584.1 | - | 3 - 4 | 547 | 61 | |
BC127117.1 | - | 2 - 3 | 202 | 61 | |
DQ068177.1 | - | 2 - 3 | 283 | 61 |