Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRIP2
Entrez Gene ID
83714
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.12: 2825348 - 2835536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031474.2 NP_113662.1 5 - 6 830 97
XM_005253797.3 XP_005253854.1 5 - 6 981 97
XM_011521031.2 XP_011519333.1 6 - 7 772 97
GenBank mRNA AK054740.1 - 1 - 2 41 97
AK098063.1 - 5 - 6 801 97
AL136557.1 - 5 - 6 798 97
BC036063.1 - 5 - 6 799 97
BC131562.1 - 5 - 6 787 97

View Full Product Details