The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002416.2 | NP_002407.1 | 2 - 3 | 268 | 79 |
GenBank mRNA | AK311852.1 | - | 2 - 3 | 254 | 79 |
BC025745.1 | - | 2 - 3 | 243 | 79 | |
BC042178.1 | - | 2 - 3 | 216 | 79 | |
BC063122.1 | - | 2 - 3 | 259 | 79 | |
BC095396.1 | - | 2 - 3 | 254 | 79 | |
DB141449.1 | - | 2 - 3 | 270 | 79 | |
X72755.1 | - | 2 - 3 | 233 | 79 |