Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADPGK
Entrez Gene ID
83440
Gene Name
ADP dependent glucokinase
Gene Aliases
2610017G09Rik, ADP-GK
Chromosome Location
Chr.15: 72751367 - 72783785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL136873.1 - 2 - 3 198 91

View Full Product Details