The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018423.2 | NP_060893.2 | 8 - 9 | 1449 | 68 |
XM_005253417.2 | XP_005253474.1 | 7 - 8 | 1349 | 68 | |
XM_011520736.1 | XP_011519038.1 | 9 - 10 | 1252 | 68 | |
XM_011520737.2 | XP_011519039.1 | 8 - 9 | 1289 | 68 | |
XM_011520738.2 | XP_011519040.1 | 8 - 9 | 1444 | 68 | |
XM_017019579.1 | XP_016875068.1 | 8 - 9 | 1232 | 68 | |
GenBank mRNA | AK313906.1 | - | 8 - 9 | 1184 | 68 |
AL353940.1 | - | 8 - 9 | 1197 | 68 | |
BC093822.1 | - | 6 - 7 | 950 | 68 | |
BC093824.1 | - | 6 - 7 | 950 | 68 | |
KP729000.1 | - | 6 - 7 | 928 | 68 |