The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018423.2 | NP_060893.2 | 9 - 10 | 1485 | 85 |
XM_005253417.2 | XP_005253474.1 | 8 - 9 | 1385 | 85 | |
XM_011520736.1 | XP_011519038.1 | 10 - 11 | 1288 | 85 | |
XM_011520737.2 | XP_011519039.1 | 9 - 10 | 1325 | 85 | |
XM_011520738.2 | XP_011519040.1 | 9 - 10 | 1480 | 85 | |
XM_017019579.1 | XP_016875068.1 | 9 - 10 | 1268 | 85 | |
GenBank mRNA | AF251059.1 | - | 9 - 10 | 1485 | 85 |
AK313906.1 | - | 9 - 10 | 1220 | 85 | |
AL353940.1 | - | 9 - 10 | 1233 | 85 | |
AY563054.1 | - | 7 - 8 | 964 | 85 | |
BC093822.1 | - | 7 - 8 | 986 | 85 | |
BC093824.1 | - | 7 - 8 | 986 | 85 | |
KP729000.1 | - | 7 - 8 | 964 | 85 |