The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003712.1 | NP_001003712.1 | 3 - 4 | 647 | 147 |
NM_001319652.1 | NP_001306581.1 | 6 - 7 | 772 | 147 | |
NM_001319653.1 | NP_001306582.1 | 4 - 5 | 684 | 147 | |
NM_020841.4 | NP_065892.1 | 4 - 5 | 693 | 147 | |
XM_005268621.4 | XP_005268678.1 | 7 - 8 | 791 | 147 | |
XM_006719224.2 | XP_006719287.1 | 3 - 4 | 652 | 147 | |
XM_011537855.2 | XP_011536157.1 | 4 - 5 | 310 | 147 | |
XM_011537856.2 | XP_011536158.1 | 5 - 6 | 728 | 147 | |
XM_011537857.2 | XP_011536159.1 | 7 - 8 | 782 | 147 | |
XM_017018768.1 | XP_016874257.1 | 4 - 5 | 325 | 147 | |
XM_017018769.1 | XP_016874258.1 | 5 - 6 | 696 | 147 | |
GenBank mRNA | AB040884.3 | - | 4 - 5 | 364 | 147 |
AF323730.1 | - | 3 - 4 | 647 | 147 | |
AF392452.1 | - | 4 - 5 | 694 | 147 | |
AK055835.1 | - | 3 - 4 | 245 | 147 | |
AK289997.1 | - | 4 - 5 | 413 | 147 | |
BC082235.1 | - | 4 - 5 | 632 | 147 | |
BC093834.1 | - | 3 - 4 | 292 | 147 | |
BC101529.1 | - | 3 - 4 | 292 | 147 | |
BC111728.1 | - | 4 - 5 | 587 | 147 | |
BX647211.1 | - | 3 - 4 | 246 | 147 | |
CR749542.1 | - | 6 - 7 | 498 | 147 |