The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003712.1 | NP_001003712.1 | 8 - 9 | 1194 | 93 |
NM_001319652.1 | NP_001306581.1 | 11 - 12 | 1319 | 93 | |
NM_001319653.1 | NP_001306582.1 | 9 - 10 | 1231 | 93 | |
NM_001319655.1 | NP_001306584.1 | 6 - 7 | 1125 | 93 | |
NM_020841.4 | NP_065892.1 | 9 - 10 | 1240 | 93 | |
XM_005268621.4 | XP_005268678.1 | 12 - 13 | 1338 | 93 | |
XM_006719224.2 | XP_006719287.1 | 8 - 9 | 1199 | 93 | |
XM_011537855.2 | XP_011536157.1 | 9 - 10 | 857 | 93 | |
XM_011537856.2 | XP_011536158.1 | 10 - 11 | 1275 | 93 | |
XM_011537857.2 | XP_011536159.1 | 12 - 13 | 1329 | 93 | |
XM_017018768.1 | XP_016874257.1 | 9 - 10 | 872 | 93 | |
XM_017018769.1 | XP_016874258.1 | 10 - 11 | 1243 | 93 | |
GenBank mRNA | AB040884.3 | - | 9 - 10 | 911 | 93 |
AF323730.1 | - | 8 - 9 | 1194 | 93 | |
AF392452.1 | - | 9 - 10 | 1241 | 93 | |
AK055835.1 | - | 8 - 9 | 792 | 93 | |
AK092160.1 | - | 6 - 7 | 1126 | 93 | |
AK289997.1 | - | 9 - 10 | 960 | 93 | |
BC082235.1 | - | 9 - 10 | 1179 | 93 | |
BC093834.1 | - | 8 - 9 | 839 | 93 | |
BC101529.1 | - | 8 - 9 | 839 | 93 | |
BC111728.1 | - | 9 - 10 | 1134 | 93 | |
CR749542.1 | - | 11 - 12 | 1044 | 93 |