Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
-
Chromosome Location
Chr.5: 66596348 - 67169595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164664.1 NP_001158136.1 8 - 9 1374 75
NM_001290226.1 NP_001277155.1 6 - 7 826 75
NM_001290227.1 NP_001277156.1 6 - 7 598 75
NM_001297651.1 NP_001284580.1 5 - 6 858 75
NM_015183.2 NP_055998.1 7 - 8 567 75
XM_006714606.3 XP_006714669.1 6 - 7 820 75
XM_006714610.2 XP_006714673.1 5 - 6 807 75
XM_011543382.2 XP_011541684.1 10 - 11 1466 75
XM_011543384.2 XP_011541686.1 5 - 6 2525 75
XM_011543385.2 XP_011541687.1 5 - 6 3645 75
XM_011543386.2 XP_011541688.1 5 - 6 476 75
XM_017009447.1 XP_016864936.1 11 - 12 1547 75
XM_017009448.1 XP_016864937.1 9 - 10 1430 75
XM_017009449.1 XP_016864938.1 6 - 7 865 75
XM_017009450.1 XP_016864939.1 6 - 7 559 75
XM_017009451.1 XP_016864940.1 6 - 7 559 75
XM_017009452.1 XP_016864941.1 6 - 7 3874 75
XM_017009453.1 XP_016864942.1 5 - 6 10689 75
GenBank mRNA AK057601.1 - 5 - 6 811 75
AK131421.1 - 7 - 8 908 75
AK296707.1 - 6 - 7 563 75
AK299538.1 - 6 - 7 708 75
AK299706.1 - 5 - 6 857 75
AY830839.1 - 7 - 8 568 75

View Full Product Details