The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000264.3 | NP_000255.2 | 2 - 3 | 584 | 129 |
NM_001083602.1 | NP_001077071.1 | 2 - 3 | 692 | 129 | |
NM_001083603.1 | NP_001077072.1 | 2 - 3 | 538 | 129 | |
NM_001083604.1 | NP_001077073.1 | 2 - 3 | 302 | 129 | |
NM_001083605.1 | NP_001077074.1 | 2 - 3 | 285 | 129 | |
NM_001083606.1 | NP_001077075.1 | 2 - 3 | 384 | 129 | |
NM_001083607.1 | NP_001077076.1 | 2 - 3 | 458 | 129 | |
GenBank mRNA | AB189436.1 | - | 2 - 3 | 692 | 129 |
AB189437.1 | - | 2 - 3 | 538 | 129 | |
AB189438.1 | - | 2 - 3 | 304 | 129 | |
AB189439.1 | - | 2 - 3 | 287 | 129 | |
AB189440.1 | - | 2 - 3 | 457 | 129 | |
AB209495.1 | - | 1 - 2 | 195 | 129 | |
AB233424.1 | - | 1 - 2 | 199 | 129 | |
BC043542.1 | - | 2 - 3 | 784 | 129 | |
U43148.1 | - | 2 - 3 | 384 | 129 | |
U59464.1 | - | 2 - 3 | 396 | 129 |