The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038707.1 | NP_001033796.1 | 1 - 2 | 169 | 67 |
XM_017001847.1 | XP_016857336.1 | 1 - 2 | 118 | 67 | |
GenBank mRNA | BX339337.2 | - | 1 - 2 | 82 | 67 |