The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282606.1 | NP_001269535.1 | 5 - 6 | 604 | 157 |
NM_001282609.1 | NP_001269538.1 | 4 - 5 | 463 | 157 | |
NM_001282610.1 | NP_001269539.1 | 4 - 5 | 425 | 157 | |
NM_016056.3 | NP_057140.2 | 4 - 5 | 463 | 157 | |
GenBank mRNA | AF113127.1 | - | 4 - 5 | 385 | 157 |
AF151877.1 | - | 4 - 5 | 402 | 157 | |
AF161526.1 | - | 4 - 5 | 354 | 157 | |
AF182041.1 | - | 4 - 5 | 390 | 157 | |
AF182414.1 | - | 5 - 6 | 491 | 157 | |
AK075069.1 | - | 4 - 5 | 377 | 157 | |
AK225158.1 | - | 4 - 5 | 383 | 157 | |
AL117550.1 | - | 4 - 5 | 842 | 157 | |
BC004401.1 | - | 4 - 5 | 380 | 157 | |
BC020613.1 | - | 4 - 5 | 366 | 157 | |
BC052273.1 | - | 4 - 5 | 389 | 157 | |
BG534013.1 | - | 4 - 5 | 420 | 158 |