The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282606.1 | NP_001269535.1 | 7 - 8 | 767 | 86 |
NM_001282610.1 | NP_001269539.1 | 6 - 7 | 588 | 86 | |
NM_016056.3 | NP_057140.2 | 6 - 7 | 626 | 86 | |
GenBank mRNA | AF113127.1 | - | 6 - 7 | 548 | 86 |
AF151877.1 | - | 6 - 7 | 565 | 86 | |
AF182041.1 | - | 6 - 7 | 553 | 86 | |
AK075069.1 | - | 6 - 7 | 540 | 86 | |
AK225158.1 | - | 6 - 7 | 546 | 86 | |
AL117550.1 | - | 6 - 7 | 1005 | 86 | |
BC004401.1 | - | 6 - 7 | 543 | 86 | |
BC020613.1 | - | 6 - 7 | 529 | 85 |