The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006289.3 | NP_006280.3 | 51 - 52 | 7222 | 96 |
GenBank mRNA | AB028950.2 | - | 51 - 52 | 7197 | 96 |
AF078828.1 | - | 51 - 52 | 6994 | 96 | |
AF177198.1 | - | 51 - 52 | 6958 | 96 | |
BC042923.1 | - | 51 - 52 | 7008 | 96 |