The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052924.2 | NP_443156.2 | 7 - 8 | 887 | 75 |
XM_005250772.3 | XP_005250829.1 | 8 - 9 | 1073 | 75 | |
XM_005250773.3 | XP_005250830.1 | 8 - 9 | 1073 | 75 | |
XM_006716502.3 | XP_006716565.1 | 8 - 9 | 1091 | 75 | |
XM_011516817.2 | XP_011515119.1 | 8 - 9 | 1091 | 75 | |
XM_011516818.1 | XP_011515120.1 | 8 - 9 | 1748 | 75 | |
XM_011516819.2 | XP_011515121.1 | 7 - 8 | 932 | 75 | |
XM_011516820.2 | XP_011515122.1 | 7 - 8 | 1275 | 75 | |
XM_011516821.2 | XP_011515123.1 | 7 - 8 | 1279 | 75 | |
XM_011516822.2 | XP_011515124.1 | 7 - 8 | 1424 | 75 | |
XM_011516823.2 | XP_011515125.1 | 8 - 9 | 1092 | 75 | |
XM_017013013.1 | XP_016868502.1 | 7 - 8 | 963 | 75 | |
XM_017013014.1 | XP_016868503.1 | 8 - 9 | 1092 | 75 | |
XM_017013015.1 | XP_016868504.1 | 7 - 8 | 1405 | 75 | |
XM_017013016.1 | XP_016868505.1 | 8 - 9 | 1073 | 75 | |
GenBank mRNA | AB067516.1 | - | 7 - 8 | 745 | 75 |
AY082588.1 | - | 7 - 8 | 754 | 75 | |
BC025767.1 | - | 7 - 8 | 855 | 75 | |
BX341691.2 | - | 7 - 8 | 891 | 74 |