Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALG13
Entrez Gene ID
79868
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location
Chr.X: 111680741 - 111764164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099922.2 NP_001093392.1 6 - 7 980 149
NM_001257230.1 NP_001244159.1 6 - 7 876 149
NM_001257231.1 NP_001244160.1 6 - 7 994 149
NM_001257234.1 NP_001244163.1 6 - 7 949 149
NM_001257237.1 NP_001244166.1 6 - 7 1053 149
NM_001324292.1 NP_001311221.1 6 - 7 980 149
NM_001324293.1 NP_001311222.1 6 - 7 1053 149
XM_006724693.2 XP_006724756.1 6 - 7 1353 149
XM_006724697.3 XP_006724760.2 6 - 7 1253 149
XM_006724698.3 XP_006724761.2 6 - 7 1378 149
XM_011531028.2 XP_011529330.1 6 - 7 1354 149
XM_011531030.1 XP_011529332.1 6 - 7 1352 149
XM_011531031.1 XP_011529333.1 6 - 7 1053 149
XM_011531032.1 XP_011529334.1 6 - 7 859 149
XM_011531033.2 XP_011529335.1 6 - 7 899 149
XM_011531034.2 XP_011529336.1 4 - 5 593 149
XM_017029846.1 XP_016885335.1 6 - 7 1355 149
XM_017029847.1 XP_016885336.1 6 - 7 1354 149
XM_017029848.1 XP_016885337.1 6 - 7 1352 149
XM_017029849.1 XP_016885338.1 6 - 7 1353 149
XM_017029850.1 XP_016885339.1 6 - 7 1352 149
XM_017029851.1 XP_016885340.1 7 - 8 1261 149
XM_017029853.1 XP_016885342.1 3 - 4 277 149
GenBank mRNA AK300394.1 - 6 - 7 818 149
AK302729.1 - 6 - 7 921 149
AK302890.1 - 6 - 7 897 149
AK304712.1 - 6 - 7 870 149
AK316306.1 - 6 - 7 922 149
AK316522.1 - 6 - 7 997 149

View Full Product Details