Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53A
Entrez Gene ID
152877
Gene Name
family with sequence similarity 53 member A
Gene Aliases
DNTNP
Chromosome Location
Chr.4: 1574055 - 1684313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297435.1 NP_001284364.1 1 - 2 431 67

View Full Product Details