The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001121.3 | NP_001112.2 | 10 - 11 | 1791 | 157 |
NM_001320591.1 | NP_001307520.1 | 11 - 12 | 1712 | 157 | |
NM_001320592.1 | NP_001307521.1 | 10 - 11 | 1775 | 157 | |
NM_001320593.1 | NP_001307522.1 | 10 - 11 | 1771 | 157 | |
NM_001320594.1 | NP_001307523.1 | 11 - 12 | 1845 | 157 | |
NM_016824.4 | NP_058432.1 | 10 - 11 | 1791 | 157 | |
NM_019903.4 | NP_063968.1 | 10 - 11 | 1775 | 157 | |
GenBank mRNA | AB209810.1 | - | 10 - 11 | 1554 | 157 |
AK223270.1 | - | 11 - 12 | 1381 | 157 | |
AK291488.1 | - | 10 - 11 | 1607 | 157 | |
BC043143.1 | - | 10 - 11 | 1516 | 157 | |
BC057285.1 | - | 10 - 11 | 1501 | 157 | |
BC062559.1 | - | 10 - 11 | 1764 | 157 | |
BX647403.1 | - | 10 - 11 | 1774 | 157 | |
BX647442.1 | - | 1 - 2 | 1252 | 157 | |
BX648016.1 | - | 10 - 11 | 1744 | 157 | |
D67031.1 | - | 10 - 11 | 1581 | 157 | |
EF553523.1 | - | 11 - 12 | 1713 | 157 | |
U37122.1 | - | 10 - 11 | 1533 | 157 |