Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSR2
Entrez Gene ID
90121
Gene Name
TSR2, ribosome maturation factor
Gene Aliases
DBA14, DT1P1A10, WGG1
Chromosome Location
Chr.X: 54440420 - 54445297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058163.1 NP_477511.1 1 - 2 84 121
GenBank mRNA BC007699.1 - 1 - 2 84 121
BC011825.1 - 1 - 2 84 121

View Full Product Details