The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324444.1 | NP_001311373.1 | 6 - 7 | 648 | 93 |
NM_001324445.1 | NP_001311374.1 | 5 - 6 | 1363 | 93 | |
NM_001324446.1 | NP_001311375.1 | 6 - 7 | 1381 | 93 | |
NM_001324448.1 | NP_001311377.1 | 6 - 7 | 590 | 93 | |
NM_001324449.1 | NP_001311378.1 | 5 - 6 | 1363 | 93 | |
NM_001324450.1 | NP_001311379.1 | 6 - 7 | 648 | 93 | |
NM_001324451.1 | NP_001311380.1 | 5 - 6 | 1421 | 93 | |
NM_001324452.1 | NP_001311381.1 | 4 - 5 | 1308 | 93 | |
NM_001324453.1 | NP_001311382.1 | 4 - 5 | 1308 | 93 | |
NM_012091.4 | NP_036223.2 | 6 - 7 | 590 | 93 | |
GenBank mRNA | AF125188.1 | - | 5 - 6 | 470 | 93 |
AK296834.1 | - | 7 - 8 | 461 | 93 | |
AK314652.1 | - | 6 - 7 | 532 | 93 | |
BC002758.2 | - | 6 - 7 | 536 | 93 |