Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
Chromosome Location
Chr.22: 45502839 - 45601134 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001996.3 NP_001987.2 10 - 11 1340 98
NM_006485.3 NP_006476.2 10 - 11 1340 98
NM_006486.2 NP_006477.2 10 - 11 1340 98
NM_006487.2 NP_006478.2 10 - 11 1340 98
GenBank mRNA AF126110.1 - 10 - 11 1203 98
AF217999.1 - 10 - 11 1283 98
AK075460.1 - 11 - 12 1410 98
AK075566.1 - 10 - 11 1192 98
AK300806.1 - 10 - 11 1276 98
AK316291.1 - 10 - 11 1276 98
BC022497.1 - 10 - 11 1296 98
U01244.1 - 10 - 11 1203 98
X53741.1 - 10 - 11 1203 98
X53742.1 - 10 - 11 1203 98
X53743.1 - 10 - 11 1203 98

View Full Product Details