Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
Chromosome Location
Chr.22: 45502839 - 45601134 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001996.3 NP_001987.2 7 - 8 929 74
NM_006485.3 NP_006476.2 7 - 8 929 74
NM_006486.2 NP_006477.2 7 - 8 929 74
NM_006487.2 NP_006478.2 7 - 8 929 74
GenBank mRNA AF126110.1 - 7 - 8 792 74
AF217999.1 - 7 - 8 872 74
AK075460.1 - 8 - 9 999 74
AK075566.1 - 7 - 8 781 74
AK300806.1 - 7 - 8 865 74
AK316291.1 - 7 - 8 865 74
BC022497.1 - 7 - 8 885 74
U01244.1 - 7 - 8 792 74
X53741.1 - 7 - 8 792 74
X53742.1 - 7 - 8 792 74
X53743.1 - 7 - 8 792 74

View Full Product Details