Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMU
Entrez Gene ID
55687
Gene Name
tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
Gene Aliases
LCAL3, MTO2, MTU1, TRMT, TRMT1, TRNT1
Chromosome Location
Chr.22: 46335401 - 46357340 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282782.1 NP_001269711.1 9 - 10 1359 135
NM_001282783.1 NP_001269712.1 9 - 10 1300 135
NM_018006.4 NP_060476.2 10 - 11 1466 135
NR_104240.1 10 - 11 1411 135
NR_104241.1 9 - 10 1304 135
XM_005261678.1 XP_005261735.1 10 - 11 1094 135
XM_011530271.2 XP_011528573.1 10 - 11 1229 135
XM_011530272.1 XP_011528574.1 9 - 10 1052 135
XM_011530274.2 XP_011528576.1 9 - 10 1066 135
XM_011530275.1 XP_011528577.1 10 - 11 1050 135
GenBank mRNA AB178028.1 - 10 - 11 1153 135
AK001002.1 - 10 - 11 1098 135
AK093743.1 - 2 - 3 2156 135
AK290712.1 - 10 - 11 1146 135
AY062123.2 - 10 - 11 1442 135
BC027991.1 - 9 - 10 958 135
DQ309998.1 - 9 - 10 1280 135
DQ309999.1 - 9 - 10 1335 135

View Full Product Details