The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282782.1 | NP_001269711.1 | 5 - 6 | 962 | 112 |
NM_001282783.1 | NP_001269712.1 | 5 - 6 | 903 | 112 | |
NM_001282784.1 | NP_001269713.1 | 5 - 6 | 903 | 112 | |
NM_001282785.1 | NP_001269714.1 | 6 - 7 | 1069 | 112 | |
NM_018006.4 | NP_060476.2 | 6 - 7 | 1069 | 112 | |
NR_104240.1 | 6 - 7 | 1014 | 112 | ||
NR_104241.1 | 5 - 6 | 907 | 112 | ||
XM_005261678.1 | XP_005261735.1 | 6 - 7 | 697 | 112 | |
XM_005261681.1 | XP_005261738.1 | 6 - 7 | 697 | 112 | |
XM_011530271.2 | XP_011528573.1 | 6 - 7 | 832 | 112 | |
XM_011530274.2 | XP_011528576.1 | 5 - 6 | 669 | 112 | |
XM_011530275.1 | XP_011528577.1 | 6 - 7 | 653 | 112 | |
GenBank mRNA | AB178028.1 | - | 6 - 7 | 756 | 112 |
AK001002.1 | - | 6 - 7 | 701 | 112 | |
AK290712.1 | - | 6 - 7 | 749 | 112 | |
AK295173.1 | - | 6 - 7 | 756 | 112 | |
AY062123.2 | - | 6 - 7 | 1045 | 112 | |
BC027991.1 | - | 5 - 6 | 561 | 112 | |
BC080631.1 | - | 5 - 6 | 565 | 112 | |
CR456445.1 | - | 6 - 7 | 725 | 112 | |
DQ309998.1 | - | 5 - 6 | 883 | 112 | |
DQ309999.1 | - | 5 - 6 | 938 | 112 | |
DQ310000.1 | - | 5 - 6 | 879 | 112 |