The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282782.1 | NP_001269711.1 | 8 - 9 | 1279 | 64 |
NM_001282783.1 | NP_001269712.1 | 8 - 9 | 1220 | 64 | |
NM_018006.4 | NP_060476.2 | 9 - 10 | 1386 | 64 | |
NR_104240.1 | 9 - 10 | 1331 | 64 | ||
NR_104241.1 | 8 - 9 | 1224 | 64 | ||
XM_005261678.1 | XP_005261735.1 | 9 - 10 | 1014 | 64 | |
XM_011530271.2 | XP_011528573.1 | 9 - 10 | 1149 | 64 | |
XM_011530272.1 | XP_011528574.1 | 8 - 9 | 972 | 64 | |
XM_011530274.2 | XP_011528576.1 | 8 - 9 | 986 | 64 | |
XM_011530275.1 | XP_011528577.1 | 9 - 10 | 970 | 64 | |
GenBank mRNA | AB178028.1 | - | 9 - 10 | 1073 | 64 |
AK001002.1 | - | 9 - 10 | 1018 | 64 | |
AK093743.1 | - | 1 - 2 | 2076 | 64 | |
AK290712.1 | - | 9 - 10 | 1066 | 64 | |
AY062123.2 | - | 9 - 10 | 1362 | 64 | |
BC027991.1 | - | 8 - 9 | 878 | 64 | |
DQ309998.1 | - | 8 - 9 | 1200 | 64 | |
DQ309999.1 | - | 8 - 9 | 1255 | 64 |