Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288814.1 NP_001275743.1 13 - 14 1461 125
NM_032450.2 NP_115826.2 14 - 15 1574 125
XM_011517270.2 XP_011515572.1 14 - 15 1712 125
XM_011517271.2 XP_011515573.1 14 - 15 1711 125
XM_011517272.2 XP_011515574.1 14 - 15 1712 125
XM_011517273.2 XP_011515575.1 14 - 15 1710 125
XM_011517274.2 XP_011515576.1 14 - 15 1710 125
XM_011517275.2 XP_011515577.1 14 - 15 1710 125
XM_011517276.2 XP_011515578.1 14 - 15 1738 125
XM_011517277.1 XP_011515579.1 14 - 15 1574 125
XM_011517278.1 XP_011515580.1 13 - 14 1454 125
XM_011517279.2 XP_011515581.1 13 - 14 1618 125
XM_011517281.1 XP_011515583.1 13 - 14 1627 125
XM_011517282.1 XP_011515584.1 12 - 13 1552 125
XM_011517283.2 XP_011515585.1 14 - 15 1533 125
XM_011517285.1 XP_011515587.1 13 - 14 1454 125
XM_017013802.1 XP_016869291.1 14 - 15 1710 125
XM_017013803.1 XP_016869292.1 14 - 15 1709 125
XM_017013804.1 XP_016869293.1 14 - 15 1574 125
XM_017013805.1 XP_016869294.1 13 - 14 1454 125
XM_017013806.1 XP_016869295.1 11 - 12 1189 125
GenBank mRNA AB058736.1 - 13 - 14 1466 125
AK092861.1 - 3 - 4 254 125
AK295740.1 - 13 - 14 1454 125
AL834309.1 - 13 - 14 1458 125

View Full Product Details