Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099280.2 NP_001092750.1 7 - 8 796 122
NM_001099281.2 NP_001092751.1 6 - 7 676 122
NM_001288814.1 NP_001275743.1 6 - 7 683 122
NM_032450.2 NP_115826.2 7 - 8 796 122
XM_011517270.2 XP_011515572.1 7 - 8 934 122
XM_011517271.2 XP_011515573.1 7 - 8 933 122
XM_011517272.2 XP_011515574.1 7 - 8 934 122
XM_011517273.2 XP_011515575.1 7 - 8 932 122
XM_011517274.2 XP_011515576.1 7 - 8 932 122
XM_011517275.2 XP_011515577.1 7 - 8 932 122
XM_011517276.2 XP_011515578.1 7 - 8 960 122
XM_011517277.1 XP_011515579.1 7 - 8 796 122
XM_011517278.1 XP_011515580.1 6 - 7 676 122
XM_011517279.2 XP_011515581.1 6 - 7 840 122
XM_011517281.1 XP_011515583.1 6 - 7 849 122
XM_011517282.1 XP_011515584.1 5 - 6 774 122
XM_011517283.2 XP_011515585.1 7 - 8 755 122
XM_011517285.1 XP_011515587.1 6 - 7 676 122
XM_011517286.2 XP_011515588.1 7 - 8 928 122
XM_017013802.1 XP_016869291.1 7 - 8 932 122
XM_017013803.1 XP_016869292.1 7 - 8 931 122
XM_017013804.1 XP_016869293.1 7 - 8 796 122
XM_017013805.1 XP_016869294.1 6 - 7 676 122
XM_017013806.1 XP_016869295.1 4 - 5 411 122
XM_017013807.1 XP_016869296.1 7 - 8 929 122
GenBank mRNA AB058736.1 - 6 - 7 688 122
AK093563.1 - 7 - 8 795 122
AK295740.1 - 6 - 7 676 122
AL834309.1 - 6 - 7 680 122
BC040742.1 - 4 - 5 423 122
BC057824.1 - 7 - 8 776 122
BC067855.1 - 7 - 8 776 122
BC092486.1 - 6 - 7 656 122

View Full Product Details