The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031282.2 | NP_112572.1 | 11 - 12 | 1597 | 75 |
XM_011510034.1 | XP_011508336.1 | 11 - 12 | 1573 | 75 | |
GenBank mRNA | AF329490.1 | - | 11 - 12 | 1461 | 75 |
AF329492.1 | - | 8 - 9 | 1420 | 75 | |
AF343659.1 | - | 11 - 12 | 1597 | 75 | |
AF343660.1 | - | 11 - 12 | 1597 | 75 | |
AF343661.1 | - | 11 - 12 | 1597 | 75 | |
AF397452.1 | - | 11 - 12 | 1461 | 75 | |
BC125173.1 | - | 11 - 12 | 1555 | 75 | |
BC125174.1 | - | 11 - 12 | 1555 | 75 | |
BC143628.1 | - | 9 - 10 | 1186 | 75 |