The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014856.2 | NP_055671.2 | 8 - 9 | 1584 | 105 |
XM_005245678.4 | XP_005245735.1 | 8 - 9 | 1741 | 105 | |
XM_005245679.2 | XP_005245736.1 | 8 - 9 | 1591 | 105 | |
XM_011510219.1 | XP_011508521.1 | 7 - 8 | 1206 | 105 | |
XM_011510220.2 | XP_011508522.1 | 7 - 8 | 1404 | 105 | |
XM_011510221.2 | XP_011508523.1 | 6 - 7 | 1112 | 105 | |
XM_011510222.1 | XP_011508524.1 | 7 - 8 | 1206 | 105 | |
XM_017002993.1 | XP_016858482.1 | 5 - 6 | 966 | 105 | |
GenBank mRNA | AB007945.2 | - | 8 - 9 | 1584 | 105 |
AK226053.1 | - | 3 - 4 | 233 | 105 |