Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECM1
Entrez Gene ID
1893
Gene Name
extracellular matrix protein 1
Gene Aliases
URBWD
Chromosome Location
Chr.1: 150508011 - 150513789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202858.1 NP_001189787.1 4 - 5 587 145
NM_004425.3 NP_004416.2 4 - 5 506 145
NM_022664.2 NP_073155.2 4 - 5 506 145
GenBank mRNA AK097046.1 - 4 - 5 522 145
AK292435.1 - 4 - 5 408 145
AK301369.1 - 4 - 5 388 145
AK302279.1 - 4 - 5 367 145
AK316390.1 - 4 - 5 367 145
BC023505.2 - 4 - 5 432 145
U65932.1 - 4 - 5 338 145
U68186.1 - 4 - 5 410 145
U68187.1 - 4 - 5 410 145

View Full Product Details