The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014077.3 | NP_054796.1 | 3 - 4 | 310 | 61 |
GenBank mRNA | AF151902.1 | - | 3 - 4 | 269 | 61 |
AK312058.1 | - | 3 - 4 | 293 | 61 | |
AL050157.1 | - | 3 - 4 | 288 | 61 | |
BC000639.1 | - | 3 - 4 | 288 | 61 | |
BC004447.1 | - | 2 - 3 | 140 | 61 | |
BC017286.1 | - | 3 - 4 | 271 | 61 | |
BG749750.1 | - | 2 - 3 | 437 | 61 |