Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAGEC3
Entrez Gene ID
139081
Gene Name
MAGE family member C3
Gene Aliases
CT7.2, HCA2, MAGE-C3, MAGEC4
Chromosome Location
Chr.X: 141838316 - 141897832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138702.1 NP_619647.1 1 - 2 125 123
GenBank mRNA AF490508.1 - 1 - 2 125 123

View Full Product Details