The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281301.1 | NP_001268230.1 | 9 - 10 | 1150 | 93 |
NM_004856.6 | NP_004847.2 | 10 - 11 | 1287 | 93 | |
NM_138555.3 | NP_612565.1 | 10 - 11 | 1287 | 93 | |
XM_005254796.2 | XP_005254853.1 | 11 - 12 | 1278 | 93 | |
XM_005254797.2 | XP_005254854.1 | 11 - 12 | 1278 | 93 | |
XM_005254798.2 | XP_005254855.1 | 11 - 12 | 1278 | 93 | |
XM_005254799.2 | XP_005254856.1 | 11 - 12 | 1278 | 93 | |
XM_011522238.1 | XP_011520540.1 | 10 - 11 | 1113 | 93 | |
XM_011522239.2 | XP_011520541.1 | 10 - 11 | 1138 | 93 | |
XM_011522240.1 | XP_011520542.1 | 2 - 3 | 132 | 93 | |
XM_017022733.1 | XP_016878222.1 | 11 - 12 | 1278 | 93 | |
GenBank mRNA | AK303874.1 | - | 6 - 7 | 681 | 93 |
AK308322.1 | - | 9 - 10 | 1072 | 93 | |
AK308361.1 | - | 4 - 5 | 496 | 93 | |
AK310402.1 | - | 12 - 13 | 1356 | 93 | |
BC017705.2 | - | 9 - 10 | 1055 | 93 | |
BC051826.1 | - | 10 - 11 | 1229 | 93 | |
BX477359.1 | - | 1 - 2 | 71 | 93 | |
X67155.2 | - | 9 - 10 | 1127 | 93 |