Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF23
Entrez Gene ID
9493
Gene Name
kinesin family member 23
Gene Aliases
CHO1, KNSL5, MKLP-1, MKLP1
Chromosome Location
Chr.15: 69414246 - 69448427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281301.1 NP_001268230.1 11 - 12 1357 67
NM_004856.6 NP_004847.2 12 - 13 1494 67
NM_138555.3 NP_612565.1 12 - 13 1494 67
XM_005254796.2 XP_005254853.1 13 - 14 1485 67
XM_005254797.2 XP_005254854.1 13 - 14 1485 67
XM_005254798.2 XP_005254855.1 13 - 14 1485 67
XM_005254799.2 XP_005254856.1 13 - 14 1485 67
XM_011522238.1 XP_011520540.1 12 - 13 1320 67
XM_011522239.2 XP_011520541.1 12 - 13 1345 67
XM_011522240.1 XP_011520542.1 4 - 5 339 67
XM_017022733.1 XP_016878222.1 13 - 14 1485 67
GenBank mRNA AK303874.1 - 8 - 9 888 67
AK308322.1 - 11 - 12 1279 67
AK308361.1 - 6 - 7 703 67
AK310402.1 - 14 - 15 1563 67
BC017705.2 - 11 - 12 1262 67
BC051826.1 - 12 - 13 1436 67
BX477359.1 - 3 - 4 278 67
X67155.2 - 11 - 12 1334 67

View Full Product Details