The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281301.1 | NP_001268230.1 | 6 - 7 | 818 | 100 |
NM_004856.6 | NP_004847.2 | 7 - 8 | 955 | 100 | |
NM_138555.3 | NP_612565.1 | 7 - 8 | 955 | 100 | |
XM_011522239.2 | XP_011520541.1 | 7 - 8 | 806 | 100 | |
GenBank mRNA | AK308322.1 | - | 6 - 7 | 740 | 100 |
AK308361.1 | - | 1 - 2 | 164 | 100 | |
BC017705.2 | - | 6 - 7 | 723 | 100 | |
BC051826.1 | - | 7 - 8 | 897 | 100 | |
X67155.2 | - | 6 - 7 | 795 | 100 |