Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C7orf26
Entrez Gene ID
79034
Gene Name
chromosome 7 open reading frame 26
Gene Aliases
-
Chromosome Location
Chr.7: 6589368 - 6608726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303039.1 NP_001289968.1 4 - 5 855 70
GenBank mRNA BC001076.2 - 4 - 5 731 70
BC005121.1 - 4 - 5 731 70

View Full Product Details