Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7orf26
Entrez Gene ID
79034
Gene Name
chromosome 7 open reading frame 26
Gene Aliases
-
Chromosome Location
Chr.7: 6589368 - 6608726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303039.1 NP_001289968.1 6 - 7 1115 76
NM_024067.3 NP_076972.2 5 - 6 1406 76
GenBank mRNA AK056149.1 - 5 - 6 1143 76
BC001076.2 - 6 - 7 991 76
BC005121.1 - 6 - 7 991 76
CA454040.1 - 2 - 3 563 76

View Full Product Details