The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016570.2 | NP_057654.2 | 12 - 13 | 1117 | 100 |
GenBank mRNA | AF183410.1 | - | 12 - 13 | 1071 | 100 |
AF216751.1 | - | 12 - 13 | 1075 | 100 | |
AF302767.1 | - | 12 - 13 | 1070 | 100 | |
AK074520.1 | - | 12 - 13 | 1092 | 100 | |
AK222799.1 | - | 12 - 13 | 1057 | 100 | |
AL834128.2 | - | 12 - 13 | 1058 | 100 | |
BC000887.1 | - | 12 - 13 | 1070 | 100 | |
BC107794.1 | - | 12 - 13 | 1074 | 100 |