The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098629.2 | NP_001092099.1 | 1 - 2 | 110 | 103 |
NM_001098630.2 | NP_001092100.1 | 1 - 2 | 110 | 103 | |
NM_001242452.2 | NP_001229381.1 | 1 - 2 | 110 | 103 | |
XM_011516161.1 | XP_011514463.1 | 1 - 2 | 125 | 103 | |
XM_011516162.1 | XP_011514464.1 | 1 - 2 | 125 | 103 | |
GenBank mRNA | AK293654.1 | - | 1 - 2 | 110 | 103 |
AY693665.1 | - | 1 - 2 | 107 | 103 | |
AY693668.1 | - | 1 - 2 | 107 | 103 | |
BU950658.1 | - | 1 - 2 | 109 | 103 | |
KP843537.1 | - | 1 - 2 | 112 | 103 | |
U51127.1 | - | 1 - 2 | 89 | 103 |