The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152759.4 | NP_689972.3 | 1 - 2 | 69 | 62 |
GenBank mRNA | AK124107.1 | - | 1 - 2 | 69 | 62 |