The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127392.2 | NP_001120864.1 | 7 - 8 | 1223 | 82 |
NM_013279.3 | NP_037411.1 | 7 - 8 | 1123 | 82 | |
XM_005274222.1 | XP_005274279.1 | 7 - 8 | 1211 | 82 | |
XM_005274223.1 | XP_005274280.1 | 7 - 8 | 1211 | 82 | |
XM_005274224.1 | XP_005274281.1 | 7 - 8 | 1211 | 82 | |
XM_005274225.1 | XP_005274282.1 | 7 - 8 | 1211 | 82 | |
XM_005274226.1 | XP_005274283.1 | 7 - 8 | 1211 | 82 | |
XM_005274227.1 | XP_005274284.1 | 7 - 8 | 1211 | 82 | |
XM_005274228.1 | XP_005274285.1 | 7 - 8 | 1058 | 82 | |
XM_011545234.2 | XP_011543536.1 | 7 - 8 | 1097 | 82 | |
GenBank mRNA | AB023171.2 | - | 7 - 8 | 1211 | 82 |
AF086762.1 | - | 7 - 8 | 1123 | 82 |