Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr.10: 22928024 - 23038527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282745.1 NP_001269674.1 18 - 19 2471 147
NM_001282747.1 NP_001269676.1 15 - 16 2003 147
NM_173081.4 NP_775104.2 18 - 19 2492 147
XM_005252380.2 XP_005252437.1 17 - 18 2280 147
XM_005252381.2 XP_005252438.1 17 - 18 2313 147
XM_011519351.2 XP_011517653.1 9 - 10 1322 147
XM_017015831.1 XP_016871320.1 17 - 18 2259 147
GenBank mRNA AK098711.1 - 8 - 9 1032 147
AK302104.1 - 15 - 16 1950 147
AK302985.1 - 18 - 19 2458 147
BC039312.1 - 18 - 19 2485 147

View Full Product Details