Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCOR
Entrez Gene ID
84458
Gene Name
ligand dependent nuclear receptor corepressor
Gene Aliases
MLR2
Chromosome Location
Chr.10: 96832250 - 96964441 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170765.1 NP_001164236.1 2 - 3 213 77
NM_001170766.1 NP_001164237.1 2 - 3 213 77
NM_032440.3 NP_115816.2 2 - 3 233 77
XM_006718033.3 XP_006718096.1 2 - 3 521 77
XM_011540277.2 XP_011538579.1 2 - 3 521 77
XM_017016785.1 XP_016872274.1 2 - 3 142
XM_017016786.1 XP_016872275.1 2 - 3 209 77
XM_017016787.1 XP_016872276.1 2 - 3 214 77
GenBank mRNA AB058698.1 - 2 - 3 146 77
AL832106.1 - 2 - 3 193 77
AL834245.2 - 2 - 3 216 77
BC053359.1 - 2 - 3 154 77
DA507116.1 - 2 - 3 233 77
DA788606.1 - 2 - 3 210 77

View Full Product Details