Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
Chromosome Location
Chr.10: 71396934 - 71815947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171930.1 NP_001165401.1 14 - 15 1843 122
NM_001171931.1 NP_001165402.1 14 - 15 1843 122
NM_022124.5 NP_071407.4 14 - 15 1843 122
XM_006717940.3 XP_006718003.1 15 - 16 1897 122
XM_006717942.3 XP_006718005.1 13 - 14 1831 122
XM_011540039.2 XP_011538341.1 15 - 16 1897 122
XM_011540042.2 XP_011538344.1 15 - 16 1897 122
XM_011540043.2 XP_011538345.1 15 - 16 1897 122
XM_011540044.2 XP_011538346.1 15 - 16 1538 122
XM_011540045.2 XP_011538347.1 15 - 16 1897 122
XM_011540046.2 XP_011538348.1 10 - 11 1332 122
XM_011540047.2 XP_011538349.1 4 - 5 632 122
XM_011540048.2 XP_011538350.1 15 - 16 1897 122
XM_011540049.2 XP_011538351.1 15 - 16 1897 122
XM_011540051.2 XP_011538353.1 15 - 16 1897 122
XM_017016499.1 XP_016871988.1 14 - 15 1843 122
XM_017016500.1 XP_016871989.1 15 - 16 1897 122
GenBank mRNA AB058715.1 - 2 - 3 228 122
AF312024.1 - 13 - 14 1453 122
AK074160.1 - 8 - 9 885 122
AY010111.2 - 13 - 14 1453 122
BC032581.1 - 14 - 15 1849 122
BC065284.1 - 14 - 15 1820 122

View Full Product Details