Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
Chromosome Location
Chr.10: 71396934 - 71815947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171933.1 NP_001165404.1 20 - 21 3109 85
NM_001171934.1 NP_001165405.1 20 - 21 3109 85
NM_001171935.1 NP_001165406.1 2 - 3 612 85
NM_001171936.1 NP_001165407.1 2 - 3 612 85
NM_022124.5 NP_071407.4 67 - 68 9902 85
XM_006717940.3 XP_006718003.1 68 - 69 9956 85
XM_006717942.3 XP_006718005.1 66 - 67 9890 85
XM_011540039.2 XP_011538341.1 67 - 68 9953 85
XM_011540042.2 XP_011538344.1 69 - 70 9866 85
XM_011540043.2 XP_011538345.1 68 - 69 9956 85
XM_011540044.2 XP_011538346.1 68 - 69 9597 85
XM_011540045.2 XP_011538347.1 68 - 69 9956 85
XM_011540046.2 XP_011538348.1 63 - 64 9391 85
XM_011540047.2 XP_011538349.1 57 - 58 8691 85
XM_011540052.2 XP_011538354.1 40 - 41 6231 85
XM_017016499.1 XP_016871988.1 67 - 68 9902 85
XM_017016500.1 XP_016871989.1 67 - 68 9830 85
XM_017016501.1 XP_016871990.1 47 - 48 7458 85
XM_017016502.1 XP_016871991.1 47 - 48 7646 85
XM_017016503.1 XP_016871992.1 37 - 38 5634 85
XM_017016504.1 XP_016871993.1 31 - 32 5419 85
XM_017016505.1 XP_016871994.1 31 - 32 5450 85
XM_017016506.1 XP_016871995.1 30 - 31 4889 85
XM_017016507.1 XP_016871996.1 24 - 25 3711 85
XM_017016508.1 XP_016871997.1 20 - 21 3164 85
GenBank mRNA AB053445.1 - 19 - 20 3048 85
AF312024.1 - 66 - 67 9512 85
AY010111.2 - 66 - 67 9512 85
AY563161.1 - 2 - 3 612 85
AY563162.1 - 2 - 3 612 85
AY563165.1 - 20 - 21 3109 85
AY563166.1 - 20 - 21 3109 85

View Full Product Details