The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016531.5 | NP_057615.3 | 2 - 3 | 387 | 83 |
GenBank mRNA | AB024523.1 | - | 2 - 3 | 110 | 83 |
AF285837.1 | - | 2 - 3 | 214 | 83 | |
AK313459.1 | - | 2 - 3 | 357 | 83 | |
BC030662.1 | - | 2 - 3 | 362 | 83 | |
BC051687.1 | - | 2 - 3 | 359 | 83 |