The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000876.3 | NP_000867.2 | 26 - 27 | 3817 | 59 |
GenBank mRNA | AB209668.1 | - | 25 - 26 | 3440 | 59 |
J03528.1 | - | 26 - 27 | 3816 | 59 | |
Y00285.1 | - | 26 - 27 | 3816 | 59 |