The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001249.3 | NP_001240.1 | 2 - 3 | 173 | 118 |
NM_001321984.1 | NP_001308913.1 | 2 - 3 | 173 | 118 | |
NM_001321985.1 | NP_001308914.1 | 2 - 3 | 173 | 118 | |
NM_001321987.1 | NP_001308916.1 | 2 - 3 | 173 | 118 | |
NM_001321988.1 | NP_001308917.1 | 2 - 3 | 949 | 118 | |
XM_006720325.3 | XP_006720388.1 | 2 - 3 | 176 | 118 | |
XM_006720326.2 | XP_006720389.1 | 2 - 3 | 176 | 118 | |
XM_017021813.1 | XP_016877302.1 | 2 - 3 | 168 | 118 | |
XM_017021815.1 | XP_016877304.1 | 2 - 3 | 168 | 118 | |
XM_017021817.1 | XP_016877306.1 | 2 - 3 | 176 | 118 | |
GenBank mRNA | AF039918.1 | - | 1 - 2 | 108 | 118 |
AY430094.1 | - | 1 - 2 | 114 | 118 | |
BC107878.1 | - | 2 - 3 | 125 | 118 |